Learn More
Hach Company Sodium/Potassium Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501997037 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Sodium/Potassium determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Sodium/Potassium level readings. Eliminate most interferences. Pack of 100 powder pillows.
Catalog No. 50-199-7037 Supplier Hach Company Supplier No. 4451569
Ionic Strength Adjustor (ISA) for Sodium/Potassium determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Sodium/Potassium level readings. Eliminate most interferences. Pack of 100 powder pillows.