Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium Tartrate Tetrahydrate, ≥99%, MP Biomedicals™

Catalog No. 021501565 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02150156-5 500 g
ICN15015601 1 kg
ICN15015605 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02150156-5 Supplier MP Biomedicals, Inc Supplier No. 02150156.5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range ≥99%
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular Weight (g/mol) 282.22
Formula Weight 282.2
Percent Purity ≥99%
Chemical Name or Material Sodium Potassium Tartrate Tetrahydrate
Show More Show Less
EINECSNumber 206-156-8
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.