Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium Tartrate, Tetrahydrate, Powder, FCC, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18606754 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-754 2.5 kg Poly Pail
18-606-755 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-754 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14022.5KG
Add to Cart
Add to Cart

P1402, 6381-59-5, C4H4KNaO6·4H2O

Spectrum™ Chemical Sodium Potassium Tartrate, Tetrahydrate, Powder, FCC is used as a sequestrant and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Alkalinity To pass test
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 99 to 102%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.