Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium propionate, 137-40-6, MFCD00002759, 100 g
Catalog No. NC0718495
Encompass
Linear Formula: CH3CH2COONa, Molecular Weight: 96.06, ≥99.0%, Synonym: Propionic acid sodium salt. Sodium propionate has been used as a supplement in transgalacto-oligosaccharides (TOS) medium for the isolation of bifidobacteria. It has been used as a supplement in SCFA (short-chain fatty acid) diet prepared for mice.
Catalog No. NC0718495 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P1880100G
Linear Formula: CH3CH2COONa, Molecular Weight: 96.06, ≥99.0%, Synonym: Propionic acid sodium salt. Sodium propionate has been used as a supplement in transgalacto-oligosaccharides (TOS) medium for the isolation of bifidobacteria. It has been used as a supplement in SCFA (short-chain fatty acid) diet prepared for mice.