Learn More
Medchemexpress LLC Sodium propionate-¹³C | 62601-06-3 | 99.6% | 100 MG
SDP
Catalog No. 5000411519
Encompass_Preferred
Sodium propionate-13C is the 13C labeled Propionate sodium, an orally active short-chain fatty acid produced by intestinal bacteria from dietary fiber. It is used in research for its various biological activities. Stable heavy isotopes like carbon-13 have been incorporated into drug molecules as tracers for quantitation during drug development, with deuteration also affecting pharmacokinetic and metabolic profiles.
  • Increases PPAR-γ
  • Inhibits NF-κB activation
  • Reduces COX-2 expression and NO production
  • Induces apoptosis and autophagy
  • Reduces HSV-1-induced keratitis
  • Has anticancer effects against glioblastoma
  • Exhibits neuroprotective, antioxidant, and anti-inflammatory activities
  • Can be used in the research of spinal cord injury and Alzheimer's disease
Catalog No. 50-004-11519 Supplier Medchemexpress LLC Supplier No. HYB1773AS100MG
Sodium propionate-13C is the 13C labeled Propionate sodium, an orally active short-chain fatty acid produced by intestinal bacteria from dietary fiber. It is used in research for its various biological activities. Stable heavy isotopes like carbon-13 have been incorporated into drug molecules as tracers for quantitation during drug development, with deuteration also affecting pharmacokinetic and metabolic profiles.
  • Increases PPAR-γ
  • Inhibits NF-κB activation
  • Reduces COX-2 expression and NO production
  • Induces apoptosis and autophagy
  • Reduces HSV-1-induced keratitis
  • Has anticancer effects against glioblastoma
  • Exhibits neuroprotective, antioxidant, and anti-inflammatory activities
  • Can be used in the research of spinal cord injury and Alzheimer's disease