Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium propionate-d3 | 1210410-35-7 | 98.38% | 50 MG
SDP

Catalog No. 5000413010
Encompass_Preferred

Sodium propionate-d3 | 1210410-35-7 | 98.38% | 50 MG

Catalog No. 50-004-13010 Supplier Medchemexpress LLC Supplier No. HYB1773AS550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium propionate-d3 | 1210410-35-7 | 98.38% | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.