Learn More
Medchemexpress LLC Sodium propionate-d3 | 1210410-35-7 | 98.4% | 100 MG
SDP
Catalog No. 5000413218
Encompass_Preferred
Sodium propionate-d3 is the deuterium labeled Propionate sodium (HY-B1773A). Sodium propionate is an orally active short-chain fatty acid produced by intestinal bacteria from the metabolism of dietary fiber. It increases PPAR-γ, inhibits NF-κB activation, reduces COX-2 expression and NO production, and induces Apoptosis and Autophagy. Sodium propionate reduces HSV-1-induced keratitis and has anticancer effects against glioblastoma. It also exhibits neuroprotective, antioxidant, and anti-inflammatory activities, and can be used in the research of spinal cord injury and Alzheimer's disease.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13218 Supplier Medchemexpress LLC Supplier No. HYB1773AS5100MG
Sodium propionate-d3 is the deuterium labeled Propionate sodium (HY-B1773A). Sodium propionate is an orally active short-chain fatty acid produced by intestinal bacteria from the metabolism of dietary fiber. It increases PPAR-γ, inhibits NF-κB activation, reduces COX-2 expression and NO production, and induces Apoptosis and Autophagy. Sodium propionate reduces HSV-1-induced keratitis and has anticancer effects against glioblastoma. It also exhibits neuroprotective, antioxidant, and anti-inflammatory activities, and can be used in the research of spinal cord injury and Alzheimer's disease.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.