Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium propionate-d3 | 1210410-35-7 | MFCD04975446 | 98.4% | 99.08 | C3H2D3NaO2 | 10 MG
SDP

Catalog No. 5000298050
Encompass_Preferred

Sodium Propionate-d3 is a deuterium-labeled form of sodium propionate intended for research use. It serves as an isotopically labeled standard or tracer in analytical, metabolic, and biochemical studies.

  • Isotopically labeled standard for mass spectrometry and tracer studies.
  • Molecular formula C3H2D3NaO2 and molecular weight 99.08.
  • CAS number 1210410-35-7 for unique substance identification.
  • Purity approximately 98.4% suitable for analytical applications.
  • Supplied as a 10 MG solid for small-scale experiments.

Catalog No. 50-002-98050 Supplier Medchemexpress LLC Supplier No. HYB1773AS510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Propionate-d3 is a deuterium-labeled form of sodium propionate intended for research use. It serves as an isotopically labeled standard or tracer in analytical, metabolic, and biochemical studies.

  • Isotopically labeled standard for mass spectrometry and tracer studies.
  • Molecular formula C3H2D3NaO2 and molecular weight 99.08.
  • CAS number 1210410-35-7 for unique substance identification.
  • Purity approximately 98.4% suitable for analytical applications.
  • Supplied as a 10 MG solid for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.