Learn More
Medchemexpress LLC Sodium propionate-d5 | 202529-18-8 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1773AS4
Encompass_Preferred
Sodium propionate-d5 is the deuterium-labeled version of Sodium propionate, which is an orally active short-chain fatty acid. It is produced by intestinal bacteria from dietary fiber metabolism. Sodium propionate increases PPAR-γ, inhibits NF-κB activation, reduces COX-2 expression and NO production, and induces Apoptosis and Autophagy. It also reduces HSV-1-induced keratitis and has anticancer effects against glioblastoma. Additionally, it exhibits neuroprotective, antioxidant, and anti-inflammatory activities, making it useful in research for spinal cord injury and Alzheimer's disease.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14380