Learn More
Medchemexpress LLC Sodium pyrophosphate 1kg
SDP
Catalog No. 5000205797
Encompass_Preferred
Sodium pyrophosphate decahydrate can be as a buffering agent a chelator and a hydrated salt nucleating agent Sodium pyrophosphate decahydrate can also be used to a raw material for synthetic laundry scent additives[1][2]
Catalog No. 50-002-05797 Supplier Medchemexpress LLC Supplier No. HYW0945101KG
Sodium pyrophosphate decahydrate can be as a buffering agent a chelator and a hydrated salt nucleating agent Sodium pyrophosphate decahydrate can also be used to a raw material for synthetic laundry scent additives[1][2]