Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pyrophosphate, Anhydrous, Powder, FCC, 95-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606767 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-767 45 kg Poly Bottle
18-606-764 500 g Poly Bottle
18-606-765 2.5 kg Poly Bottle
18-606-766 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-767 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S141945KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1419, 7722-88-5, Na4P2O7

Spectrum™ Chemical Sodium Pyrophosphate, Anhydrous, Powder, FCC it is used as a palatability enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7722-88-5
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
MDL Number MFCD00003513
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Percent Purity 95 to 100.5%
Grade FCC
Loss on Ignition 0.005
Packaging Poly Bottle
Chemical Name or Material Sodium Pyrophosphate, Anhydrous, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.