Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pyrophosphate Decahydrate, ACS Reagent, Reagents

Catalog No. 01337686
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-686 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-686 Supplier Reagents Holdings Llc Supplier No. C1324580500A5
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Pyrophosphate Decahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Pyrophosphate Decahydrate, ACS Reagent
Boiling Point 93°C
Color Colorless/White
CAS 13472-36-1
Physical Form Solid
Quantity 500 g
Molecular Formula Na4P2O7, 10H2O
Synonym Tetrasodium Diphosphate Decahydrate
Molecular Weight (g/mol) 446.0552
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.82
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.