Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium pyrophosphate tetrabasic

Catalog No. 503125689 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5689 500 g
50-312-6201 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-5689 Supplier Sigma Aldrich Supplier No. P8010500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium pyrophosphate tetrabasic (Na4P2O7) is a non-toxic and biocompatible compound used as an electroactive media for exfoliation of the surface coating. It is also be used as an additive in the food industry.Na4P2O7 can be used as an inorganic additive to improve the stability and electrochemical performance of redox flow batteries.

Specifications

CAS 7722-88-5
Quantity 500 g
Molecular Formula Na4O7P2
Linear Formula Na4P2O7
MDL Number MFCD00003513
Synonym Sodium diphosphate tetrabasic; Tetrasodium pyrophosphate
Molecular Weight (g/mol) 265.9
Percent Purity ≥95%
Density 2.53 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.