Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pyrophosphate tetrabasic decahydrate, Puriss. p.a., ≥99%, Solstice

Catalog No. 6002264 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-64 2.5 kg
60-045-817 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-022-64 Supplier Solstice Supplier No. 304112.5KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 2.5 kg
Linear Formula Na4P2O7 · 10H2O
MDL Number MFCD00149200
UN Number NONH for all modes of transport
Formula Weight 446.06g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Sodium Pyrophosphate Tetrabasic Decahydrate
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H302-H318
EINECSNumber 231-767-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.