Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Sodium Pyruvate (100X), 100 mL
SDP

Catalog No. 507517662
Encompass_Preferred

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Krebs cycle. The 100mM solution should be diluted 1:100 for most cell culture applications.

• 100mM solution

Recommendation: Protect from light. Add to cell culture media as a carbon source at 1mM or 100mg/mL.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7662 Supplier Quality Biological Inc Supplier No. 116079721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Krebs cycle. The 100mM solution should be diluted 1:100 for most cell culture applications.

• 100mM solution

Recommendation: Protect from light. Add to cell culture media as a carbon source at 1mM or 100mg/mL.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.