Learn More
Medchemexpress LLC Sodium pyruvate | 113-24-6 | MFCD00002586 | 99.9% | 110.04 | C3H3NaO3 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01591350G
Encompass_Preferred
Sodium 2-oxopropanoate (sodium pyruvate) is a three-carbon glycolysis intermediate supplied as an off-white to light-yellow solid for research use. It is used as an energy source in cell culture, as an antioxidant and H2O2 scavenger, and as an analytical standard for metabolic and biochemical studies. The material is water-soluble and includes recommended storage and handling conditions for long-term stability.
  • High purity (99.86%) solid suitable for research applications.
  • Water soluble; forms clear solutions with ultrasonic assistance.
  • Supports cellular metabolism when added to culture media.
  • Acts as an antioxidant and hydrogen peroxide scavenger in assays.
  • Supplied with storage recommendations to maintain stability.
Catalog No. 50-003-03886