Learn More
Medchemexpress LLC Sodium pyruvate-2-13C | 87976-70-3 | MFCD00084080 | 98.9% | 111.04 | C2^13CH3NaO3 | 50mg
SDP
Catalog No. 5000269637
Encompass_Preferred
Pyruvic acid-13C (sodium) is the 13C-labeled Pyruvic acid Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats
Catalog No. 50-002-69637 Supplier Medchemexpress LLC Supplier No. HYY0781S50MG
Pyruvic acid-13C (sodium) is the 13C-labeled Pyruvic acid Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats