Learn More
Medchemexpress LLC Sodium pyruvate-2-13C | 87976-70-3 | MFCD00084080 | 98.9% | 111.04 | C2-13CH3NaO3 | 5mg
SDP
Catalog No. 5000268592
Encompass_Preferred
Pyruvic acid-13C (sodium) is the 13C-labeled Pyruvic acid Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats
Catalog No. 50-002-68592 Supplier Medchemexpress LLC Supplier No. HYY0781S5MG
Pyruvic acid-13C (sodium) is the 13C-labeled Pyruvic acid Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats