Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pyruvate, MP Biomedicals™

Catalog No. MP219965405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219965405 5 g
MP219965425 25 g
MP219965480 100 g
MP219965490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219965405 Supplier MP Biomedicals, Inc Supplier No. 0219965405
Add to Cart
Add to Cart

Sodium pyruvate, White Powder

  • It is utilized as a component in culture broth and media.
  • The use of sodium pyruvate in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described.
  • A protocol that uses sodium pyruvate to establish stably transfected human B cell lines has been published.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.04
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym Pyruvic acid sodium salt, 2-Oxopropanoic acid sodium salt, α-ketopropionic acid sodium salt

Specifications

Melting Point >300°C (literature)
Color White
pH 6.54
Loss on Drying 0.0021
Quantity 5 g
Beilstein 3568341
Solubility Information Soluble in water (0.06g/mL) - Clear and Complete.
Physical Form Powder
Assay ≥98.0%
storageNote1 Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.