Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pyruvate, MP Biomedicals

Catalog No. ICN1682049 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1682049 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1682049 Supplier MP Biomedicals, Inc Supplier No. 091682049
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

100 mM solution

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Quantity 100 mL
Chemical Name or Material Sodium Pyruvate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.