Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SS139100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS139-100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SS139-100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For atomic absorption. (1mL = 1mg Na) Solute: Sodium Chloride Solvent: Distilled Water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg
Density 1.0g/cm³
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Quantity 100 mL
Grade Certified
Chemical Name or Material Sodium, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.