Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ricinolate 93.0+%, TCI America™
SDP

Catalog No. R0030500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0030500G 500 g
R003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0030500G Supplier TCI America Supplier No. R0030500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5323-95-5
Molecular Formula C18H34NaO3
Molecular Weight (g/mol) 321.457
MDL Number MFCD00050760
InChI Key ZCEUYFQZJDGDEF-DPMBMXLASA-N
Synonym Ricinolic Acid Sodium Salt, Ricinoleic Acid Sodium Salt
PubChem CID 87064429
IUPAC Name (Z,12R)-12-hydroxyoctadec-9-enoic acid;sodium
SMILES CCCCCCC(CC=CCCCCCCCC(=O)O)O.[Na]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 320.45
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ricinolate
EINECSNumber (2)-1333&(8)-0289&(9)-1676
RTECSNumber VJ3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.