Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | 98.0% | 112.06 g/mol | C3H5NaO3 | 100 MG
SDP

Catalog No. 5000209608
Encompass_Preferred

L-lactic acid sodium is the sodium salt of the naturally occurring L-enantiomer of lactic acid. It is supplied as a water-soluble research reagent used as a biochemical building block and intermediate in synthesis.

  • Chemical formula: C3H5NaO3
  • Molecular weight: 112.06 g/mol
  • Purity: 98.0%
  • Physical form: white solid, soluble in water
  • Solubility: miscible or highly soluble in water
  • Applications: biochemical reagent, drug intermediate, and synthetic building block
  • Typical pack sizes: available in small research quantities such as 100 mg

Catalog No. 50-002-09608 Supplier Medchemexpress LLC Supplier No. HYW040233100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-lactic acid sodium is the sodium salt of the naturally occurring L-enantiomer of lactic acid. It is supplied as a water-soluble research reagent used as a biochemical building block and intermediate in synthesis.

  • Chemical formula: C3H5NaO3
  • Molecular weight: 112.06 g/mol
  • Purity: 98.0%
  • Physical form: white solid, soluble in water
  • Solubility: miscible or highly soluble in water
  • Applications: biochemical reagent, drug intermediate, and synthetic building block
  • Typical pack sizes: available in small research quantities such as 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.