Learn More
Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | ≥98.0% | 112.06 g/mol | C3H5NaO3 | 50 MG
SDP
Catalog No. 5000208614
Encompass_Preferred
Sodium L-lactate (sodium (S)-2-hydroxypropanoate) is the monosodium salt of L-lactic acid supplied as a research-grade chemical reagent. It is used as a building block in synthesis, for biochemical preparations, and as a precursor for polylactic acid production.
  • High purity (≥98.0%) suitable for research use.
  • Molecular weight 112.06 g/mol; chemical formula C3H5NaO3.
  • Water soluble; commonly handled in aqueous solutions or as the crystalline salt.
  • Supplied in small reagent quantities, for example 50 MG vials for laboratory use.
  • Used as a reagent and intermediate in polymer synthesis and biochemical assays.
Catalog No. 50-002-08614 Supplier Medchemexpress LLC Supplier No. HYW04023350MG
Sodium L-lactate (sodium (S)-2-hydroxypropanoate) is the monosodium salt of L-lactic acid supplied as a research-grade chemical reagent. It is used as a building block in synthesis, for biochemical preparations, and as a precursor for polylactic acid production.
  • High purity (≥98.0%) suitable for research use.
  • Molecular weight 112.06 g/mol; chemical formula C3H5NaO3.
  • Water soluble; commonly handled in aqueous solutions or as the crystalline salt.
  • Supplied in small reagent quantities, for example 50 MG vials for laboratory use.
  • Used as a reagent and intermediate in polymer synthesis and biochemical assays.