Learn More
Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | 99.7% | 112.06 g/mol | C3H5NaO3 | 25 MG
SDP
Catalog No. 5000209200
Encompass_Preferred
Sodium (S)-2-hydroxypropanoate is the sodium salt of L-lactic acid supplied as an analytical standard for research and analytical applications. It is provided in small, accurately weighed quantities for use as reference material, assay calibration, and method development.
  • Provided as an analytical standard for calibration and method development.
  • High purity suitable for analytical use (99.7%).
  • Molecular weight 112.06 g/mol, chemical formula C3H5NaO3.
  • Supplied in small, accurately weighed packages for precise dosing.
Catalog No. 50-002-09200 Supplier Medchemexpress LLC Supplier No. 25MGHYW040233R
Sodium (S)-2-hydroxypropanoate is the sodium salt of L-lactic acid supplied as an analytical standard for research and analytical applications. It is provided in small, accurately weighed quantities for use as reference material, assay calibration, and method development.
  • Provided as an analytical standard for calibration and method development.
  • High purity suitable for analytical use (99.7%).
  • Molecular weight 112.06 g/mol, chemical formula C3H5NaO3.
  • Supplied in small, accurately weighed packages for precise dosing.