Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | MFCD00066576 | >90.0% | 112.06 g/mol | C3H5NaO3 | 1 ML
SDP

Catalog No. 5000209733
Encompass_Preferred

Sodium (S)-2-hydroxypropanoate, the sodium salt of L-lactic acid, is supplied as a prepared solution for research use. It is used as a biochemical building block and as a precursor in polymer and metabolic studies.

  • Provided as a 10 mM solution in 1 mL volume
  • Purity ≥90%
  • CAS number 867-56-1
  • Molecular weight 112.06 g/mol
  • Suitable for research and synthetic applications

Catalog No. 50-002-09733 Supplier Medchemexpress LLC Supplier No. HYW040233B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium (S)-2-hydroxypropanoate, the sodium salt of L-lactic acid, is supplied as a prepared solution for research use. It is used as a biochemical building block and as a precursor in polymer and metabolic studies.

  • Provided as a 10 mM solution in 1 mL volume
  • Purity ≥90%
  • CAS number 867-56-1
  • Molecular weight 112.06 g/mol
  • Suitable for research and synthetic applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.