Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | MFCD00066576 | >=90.0% | 112.06 g/mol | C3H5NaO3 | 100 MG
SDP

Catalog No. 5000208458
Encompass_Preferred

Sodium (S)-2-hydroxypropanoate (sodium L-lactate) is the sodium salt of L-lactic acid supplied as a biochemical building block with a stated purity of ≥90%. It is used as a reagent and as a precursor in the production of the bioplastic polymer poly-lactic acid (PLA).

  • Sodium salt of L-lactic acid
  • Purity ≥90%
  • Molecular formula C3H5NaO3, molecular weight 112.06 g/mol
  • Used as a building block and precursor to poly-lactic acid (PLA)
  • CAS number 867-56-1

Catalog No. 50-002-08458 Supplier Medchemexpress LLC Supplier No. HYW040233B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium (S)-2-hydroxypropanoate (sodium L-lactate) is the sodium salt of L-lactic acid supplied as a biochemical building block with a stated purity of ≥90%. It is used as a reagent and as a precursor in the production of the bioplastic polymer poly-lactic acid (PLA).

  • Sodium salt of L-lactic acid
  • Purity ≥90%
  • Molecular formula C3H5NaO3, molecular weight 112.06 g/mol
  • Used as a building block and precursor to poly-lactic acid (PLA)
  • CAS number 867-56-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.