Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | MFCD00066576 | >90.0% | 112.06 g/mol | C3H5NaO3 | 50 MG
SDP

Catalog No. 5000209120
Encompass_Preferred

L-lactic acid sodium is the sodium salt of L-lactic acid provided for research and analytical use. It is used as a building block and precursor for the synthesis of polylactic acid and may exhibit biological activity in some research contexts.

  • contains sodium salt of l-lactic acid for synthesis applications.
  • provided in small research pack sizes suitable for analytical work.
  • listed cas number and formula facilitate substance identification and regulatory documentation.
  • suitable as a precursor for bioplastic production and related research.
  • storage recommended at 4 °C in sealed container per batch documentation.

Catalog No. 50-002-09120 Supplier Medchemexpress LLC Supplier No. HYW040233B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-lactic acid sodium is the sodium salt of L-lactic acid provided for research and analytical use. It is used as a building block and precursor for the synthesis of polylactic acid and may exhibit biological activity in some research contexts.

  • contains sodium salt of l-lactic acid for synthesis applications.
  • provided in small research pack sizes suitable for analytical work.
  • listed cas number and formula facilitate substance identification and regulatory documentation.
  • suitable as a precursor for bioplastic production and related research.
  • storage recommended at 4 °C in sealed container per batch documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.