Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | MFCD00066576 | ≥98.0% | 112.06 g/mol | C3H5NaO3 | 100 MG
SDP

Catalog No. 5000209388
Encompass_Preferred

L-lactic acid sodium is the sodium salt of L-lactic acid supplied as an analytical standard for research and reference use. It is provided as a white to off-white solid and is used as a reference material in analytical assays and as a building block in biochemical workflows.

  • chemical name: sodium (S)-2-hydroxypropanoate
  • cas number: 867-56-1
  • chemical formula: C3H5NaO3
  • molecular weight: 112.06 g/mol
  • purity: ≥98.0% (nmr)
  • product type: reference standard for analytical and research applications
  • packaging: supplied as a 100 mg solid

Catalog No. 50-002-09388 Supplier Medchemexpress LLC Supplier No. HYW040233R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-lactic acid sodium is the sodium salt of L-lactic acid supplied as an analytical standard for research and reference use. It is provided as a white to off-white solid and is used as a reference material in analytical assays and as a building block in biochemical workflows.

  • chemical name: sodium (S)-2-hydroxypropanoate
  • cas number: 867-56-1
  • chemical formula: C3H5NaO3
  • molecular weight: 112.06 g/mol
  • purity: ≥98.0% (nmr)
  • product type: reference standard for analytical and research applications
  • packaging: supplied as a 100 mg solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.