Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate | 867-56-1 | MFCD00066576 | 99.7% | 112.06 g/mol | C3H5NaO3 | 10 MG
SDP

Catalog No. 5000209119
Encompass_Preferred

L-Lactic acid sodium is the sodium salt of L-lactic acid supplied as an analytical standard for research and analytical applications. It is used as a reference standard, a building block for chemical synthesis, and for analytical method development.

  • Analytical standard suitable for reference and calibration.
  • CAS number 867-56-1.
  • Chemical name: sodium (S)-2-hydroxypropanoate.
  • Molecular formula C3H5NaO3, molecular weight 112.06 g/mol.
  • High purity reported for analytical use.

Catalog No. 50-002-09119 Supplier Medchemexpress LLC Supplier No. HYW040233R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Lactic acid sodium is the sodium salt of L-lactic acid supplied as an analytical standard for research and analytical applications. It is used as a reference standard, a building block for chemical synthesis, and for analytical method development.

  • Analytical standard suitable for reference and calibration.
  • CAS number 867-56-1.
  • Chemical name: sodium (S)-2-hydroxypropanoate.
  • Molecular formula C3H5NaO3, molecular weight 112.06 g/mol.
  • High purity reported for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.