Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate-d3 | 285979-84-2 | MFCD00084178 | 98.3% | 115.08 g/mol | C3H2D3NaO3 | 5 MG
SDP

Catalog No. 5000208926
Encompass_Preferred

L-Lactic acid-d3 sodium is the deuterium-labeled sodium salt of L-lactic acid (sodium (S)-2-hydroxypropanoate-d3) supplied for research use as an isotopically labeled standard or tracer. It is provided as a solid with high labeled purity and is intended for analytical and metabolic studies.

  • Isotopically labeled (d3) sodium salt.
  • High purity suitable for research (98.3%).
  • Molecular weight 115.08 g/mol.
  • Chemical formula C3H2D3NaO3 and CAS 285979-84-2.
  • Supplied in small, accurately measured quantities for analytical use.
  • Store sealed at 4°C and protect from moisture; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-08926 Supplier Medchemexpress LLC Supplier No. HYW040233S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Lactic acid-d3 sodium is the deuterium-labeled sodium salt of L-lactic acid (sodium (S)-2-hydroxypropanoate-d3) supplied for research use as an isotopically labeled standard or tracer. It is provided as a solid with high labeled purity and is intended for analytical and metabolic studies.

  • Isotopically labeled (d3) sodium salt.
  • High purity suitable for research (98.3%).
  • Molecular weight 115.08 g/mol.
  • Chemical formula C3H2D3NaO3 and CAS 285979-84-2.
  • Supplied in small, accurately measured quantities for analytical use.
  • Store sealed at 4°C and protect from moisture; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.