Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate (sodium L-lactate) | 867-56-1 | 90.0% | 112.06 g/mol | C3H5NaO3 | 25 MG
SDP

Catalog No. 5000209595
Encompass_Preferred

Sodium (S)-2-hydroxypropanoate (sodium L-lactate) is the sodium salt of L-lactic acid, supplied as a solid used as a biochemical building block and precursor for poly-lactic acid (PLA) synthesis. Typical purity is 90%.

  • CAS number 867-56-1.
  • Chemical formula C3H5NaO3.
  • Molecular weight 112.06 g/mol.
  • Purity 90.0%.
  • Physical form white to off-white solid.
  • Common applications: precursor for PLA, reagent in chemical synthesis and research.

Catalog No. 50-002-09595 Supplier Medchemexpress LLC Supplier No. 25MGHYW040233B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium (S)-2-hydroxypropanoate (sodium L-lactate) is the sodium salt of L-lactic acid, supplied as a solid used as a biochemical building block and precursor for poly-lactic acid (PLA) synthesis. Typical purity is 90%.

  • CAS number 867-56-1.
  • Chemical formula C3H5NaO3.
  • Molecular weight 112.06 g/mol.
  • Purity 90.0%.
  • Physical form white to off-white solid.
  • Common applications: precursor for PLA, reagent in chemical synthesis and research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.