Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-3-amino-3-carboxypropanoate hydrate | 323194-76-9 | MFCD00152960 | ≥98.0% | 173.10 g/mol | C4H8NNaO5 | 500 G
SDP

Catalog No. 5000283586
Encompass_Preferred

Sodium (S)-3-amino-3-carboxypropanoate hydrate is the sodium salt monohydrate of L-aspartate supplied as a high-purity biochemical reagent for research use. It is used in biochemical and physiological assays and has documented application in cardioplegia solutions. The compound is provided as a hydrated solid with characterized molecular and analytical data to support laboratory use.

  • High purity: ≥98.0%.
  • Molecular weight 173.10 g/mol.
  • Molecular formula C4H8NNaO5.
  • Hydrate state: monohydrate (hydrated solid).
  • Available in multiple pack sizes, including 500 g.
  • Intended for research use only; consult SDS for safe handling.

Catalog No. 50-002-83586 Supplier Medchemexpress LLC Supplier No. HYW140900500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium (S)-3-amino-3-carboxypropanoate hydrate is the sodium salt monohydrate of L-aspartate supplied as a high-purity biochemical reagent for research use. It is used in biochemical and physiological assays and has documented application in cardioplegia solutions. The compound is provided as a hydrated solid with characterized molecular and analytical data to support laboratory use.

  • High purity: ≥98.0%.
  • Molecular weight 173.10 g/mol.
  • Molecular formula C4H8NNaO5.
  • Hydrate state: monohydrate (hydrated solid).
  • Available in multiple pack sizes, including 500 g.
  • Intended for research use only; consult SDS for safe handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.