Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Saccharin, MilliporeSigma™ Supelco™

Catalog No. 111015622 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5622 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5622 Supplier MilliporeSigma Supelco Supplier No. 47839
Add to Cart
Add to Cart

Sodium saccharin is a white crystalline powder, which is commonly used as a synthetic or artificial sweetener in carbonated soft drinks and beverages. It can find a wide range of applications in food processing, commodities and also feed production industries.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 82385-42-0
Physical Form Neat
Linear Formula C7H4 NNaO3 S · xH2 O
MDL Number MFCD00149605
Synonym Saccharin sodium salt hydrate; 2,3-Dihydro-3-oxobenzisosulfonazole sodium salt; 2-Sulfobenzoic acid imide sodium salt; o-Sulfobenzimide sodium salt; Saccharin soluble
Type Food Additive Standard
Quantity 1 g
Formula Weight 205.17 (anhydrous basis)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.