Learn More
Medchemexpress LLC Sodium Salicylate | 54-21-7 | 00006036 | >99.8% | 1 ML
SDP
Catalog No. 5000407193
Encompass_Preferred
Sodium Salicylate (salicylic acid sodium salt) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity, functioning independently of transcription factor (NF-κB) activation. It also acts as an S6K inhibitor and a NF-κB inhibitor, reducing inflammatory gene expression and aiding in muscle repair.
  • Inhibits COX-2 activity
  • Functions as an S6K inhibitor
  • Acts as a NF-κB inhibitor, which helps to decrease inflammatory gene expression
  • Supports repair in aged muscle
  • Inhibits prostaglandin E2 release
  • Causes a concentration-dependent inhibition of COX-2 activity
Catalog No. 50-004-07193 Supplier Medchemexpress LLC Supplier No. HYB0167A10MM/1ML
Sodium Salicylate (salicylic acid sodium salt) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity, functioning independently of transcription factor (NF-κB) activation. It also acts as an S6K inhibitor and a NF-κB inhibitor, reducing inflammatory gene expression and aiding in muscle repair.
  • Inhibits COX-2 activity
  • Functions as an S6K inhibitor
  • Acts as a NF-κB inhibitor, which helps to decrease inflammatory gene expression
  • Supports repair in aged muscle
  • Inhibits prostaglandin E2 release
  • Causes a concentration-dependent inhibition of COX-2 activity