Learn More
Medchemexpress LLC Sodium Salicylate | 54-21-7 | 00006036 | 99.99% | 500 MG
SDP
Catalog No. 5000406557
Encompass_Preferred
Sodium Salicylate is an inhibitor of cyclo-oxygenase-2 (COX-2) activity, operating independently of transcription factor (NF-κB) activation. It also functions as an S6K inhibitor. This compound is identified as an NF-κB inhibitor that reduces inflammatory gene expression and supports repair processes in aged muscle.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions as an S6K inhibitor
  • Decreases inflammatory gene expression
  • Improves repair in aged muscle
  • Lowers fasting and postprandial plasma glucose levels
  • Reduces 11β-HSD1 mRNA and enzyme activity in adipose tissue
Catalog No. 50-004-06557 Supplier Medchemexpress LLC Supplier No. HYB0167A500MG
Sodium Salicylate is an inhibitor of cyclo-oxygenase-2 (COX-2) activity, operating independently of transcription factor (NF-κB) activation. It also functions as an S6K inhibitor. This compound is identified as an NF-κB inhibitor that reduces inflammatory gene expression and supports repair processes in aged muscle.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions as an S6K inhibitor
  • Decreases inflammatory gene expression
  • Improves repair in aged muscle
  • Lowers fasting and postprandial plasma glucose levels
  • Reduces 11β-HSD1 mRNA and enzyme activity in adipose tissue