Learn More
Apexbio Technology LLC Sodium salicylate 54-21-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092955
Encompass_Preferred
Sodium salicylate (CAS 54-21-7) is a small molecule and a primary metabolite of acetylsalicylic acid It modulates cellular signaling by inhibiting the activation of nuclear factor kappa B (NF- B) a transcription factor central to inflammatory and immune responses Additionally sodium salicylate has been demonstrated to attenuate oxidative stress at the cellular level This compound is widely employed in biomedical research to investigate the regulation of inflammatory pathways and oxidative damage facilitating studies in immunology pharmacology and disease pathogenesis
Catalog No. 50-000-92955 Supplier Apexbio Technology LLC Supplier No. B20285.1
Sodium salicylate (CAS 54-21-7) is a small molecule and a primary metabolite of acetylsalicylic acid It modulates cellular signaling by inhibiting the activation of nuclear factor kappa B (NF- B) a transcription factor central to inflammatory and immune responses Additionally sodium salicylate has been demonstrated to attenuate oxidative stress at the cellular level This compound is widely employed in biomedical research to investigate the regulation of inflammatory pathways and oxidative damage facilitating studies in immunology pharmacology and disease pathogenesis