Learn More
Medchemexpress LLC Sodium Salicylate | 54-21-7 | 99.99% | 50 G
SDP
Catalog No. 5000405989
Encompass_Preferred
Sodium Salicylate is a chemical compound that inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. It also acts as an S6K inhibitor and a NF-κB inhibitor, which helps to reduce inflammatory gene expression and improve muscle repair in aged muscle.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions as an S6K inhibitor
  • Acts as an NF-κB inhibitor
  • Decreases inflammatory gene expression
  • Improves repair in aged muscle
Catalog No. 50-004-05989 Supplier Medchemexpress LLC Supplier No. HYB0167A50G
Sodium Salicylate is a chemical compound that inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. It also acts as an S6K inhibitor and a NF-κB inhibitor, which helps to reduce inflammatory gene expression and improve muscle repair in aged muscle.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions as an S6K inhibitor
  • Acts as an NF-κB inhibitor
  • Decreases inflammatory gene expression
  • Improves repair in aged muscle