Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Salicylate, Reagent Grade, Reagents

Catalog No. 01337683
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-683 500 g
01-337-685 125 g
01-337-684 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-683 Supplier Reagents Holdings Llc Supplier No. C1324700500A5
Add to Cart
Add to Cart

Sodium Salicylate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym Sodium 2-hydroxybenzoate
IUPAC Name sodium 2-hydroxybenzoate
SMILES [Na+].OC1=CC=CC=C1C([O-])=O

Specifications

Boiling Point 336°C
Color White or Slightly Yellow
Grade Reagent
Quantity 500 g
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 0.32
Chemical Name or Material Sodium Salicylate, Reagent Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.