Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenate, Anhydrous, Crystal, 97%, Spectrum™ Chemical
SDP

Catalog No. 18606783 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-783 25 kg Amber Glass Bottle
18-606-779 25 g Amber Glass Bottle
18-606-780 100 g Amber Glass Bottle
18-606-781 500 g Amber Glass Bottle
18-606-782 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-783 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S142825KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

S1428, 13410-01-0, Na2SeO4

Spectrum™ Chemical Sodium Selenate, Anhydrous, Crystal has several uses, including the manufacturing of glass, specifically red-hued glass, but can also produce clear glass when mixed with ferrous oxides. It's a white or gray solid substance that is highly toxic. Regardless of the toxicity, sodium selenate is used in vitamin supplements as a trace mineral additive and is being investigated as a potential anti-Alzheimers drug candidate.

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
IUPAC Name disodium selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 kg
Percent Purity 97%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Selenate, Anhydrous, Crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.