Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenite, 99.8+%, Powder, Spectrum™ Chemical
SDP

Catalog No. 18613847 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Lined Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-847 500 g Amber Glass Bottle
18-613-849 25 g Amber Glass Bottle
18-613-846 100 g Amber Glass Bottle
18-613-850 5 kg Poly Pail
18-613-848 25 kg Lined Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-613-847 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO206500GM
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
IUPAC Name disodium selenite
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity ≥99.8%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.