Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Silicate Solution (40° -42° Be/Technical), Fisher Chemical™

Catalog No. SS3384 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS338-4 4 L Glass Bottle
SS338-1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SS338-4

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1344-09-8 , 7732-18-5
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium oxosilanebis(olate) pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Color Colorless
Boiling Point 100.6°C
Melting Point -3°C
UN Number 3266
Density 1.4g/cm³
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Specific Gravity 1.38 to 1.42 (at 25°C)
Viscosity 20-400 mPa/s at 20°C
pH 11.2
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium Silicate Solution, 40°-42° Bé
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.