Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium silicate solution, Reagent Grade, Solstice

Catalog No. 60048582 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-582 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-582 Supplier Solstice Supplier No. 3384431L
Add to Cart
Add to Cart
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 1344-09-8
Molecular Formula Na2SiO3 in H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium trisilicate solution, Water glass
SMILES O

Specifications

Boiling Point 100°C
CAS Min % 50.00, 30.00
pH 11 - 13
CAS Max % 70.00, 50.00
Physical Form Liquid
Quantity 1 L
Flash Point Not applicable
Grade Reagent
Concentration or Composition (by Analyte or Components) Water 50-70%, Sodium Silicate (MV > 3.2) 30-50%
Packaging Plastic Bottle
Hazard Category Danger
Hazard Statement H302+H314
Precautionary Statement P280+P270+P264
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.