Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Silicotungstate, MP Biomedicals

Catalog No. ICN224517 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224517 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224517 Supplier MP Biomedicals, Inc Supplier No. 0522451710

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12027-47-3
Molecular Formula H4Na4O40SiW12
Molecular Weight (g/mol) 2970.116
InChI Key RFKXKZJSLAFLCE-UHFFFAOYSA-N
Synonym sodium silicotungstate, 4na.o40siw12, tetrasodium 5-1,13-dioxido-7-5-oxido-1,1,3,3,5,5-hexaoxotritungstoxan-1-yl oxy-1,1,3,3,5,5,9,9,11,11,13,13-dodecaoxo-2,4,6,8,10,12-hexaoxa-7-sila-1,3,5,9,11,13-hexatungstatridecan-7-yl oxy-3,3,5,5-tetraoxotritungstoxan-1-oylolate
PubChem CID 25113564
IUPAC Name tetrasodium;dioxotungsteniooxy-(oxido(dioxo)tungstenio)oxy-dioxotungsten;silicic acid
SMILES O[Si](O)(O)O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 10 g
Formula Weight 2956.37
Chemical Name or Material Sodium Silicotungstate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.