Learn More
Perkin Elmer US LLC Sodium Spectral Lamp - 20 W
Catalog No. 502113176
Encompass_Preferred
This replacement lamp is for PerkinElmer's older Polarimeter models. We recommend a service visit to replace and align this lamps, to ensure optimal operation.
Catalog No. 50-211-3176 Supplier Perkin Elmer US LLC Supplier No. L9004741
This replacement lamp is for PerkinElmer's older Polarimeter models. We recommend a service visit to replace and align this lamps, to ensure optimal operation.