Learn More
Waltron LLC Sodium Low Calibration Standard, 100Ppb, 5 Gal/20L Cube
SDP
Catalog No. NC1219156
Encompass
100 ppb Sodium calibration standard for brands of sodium analyzers. Bulk packed in a 5 gallon/20 liter cube for refilling calibration solution bottles.
Catalog No. NC1219156 Supplier Waltron LLC Supplier No. N1234545
100 ppb Sodium calibration standard for brands of sodium analyzers. Bulk packed in a 5 gallon/20 liter cube for refilling calibration solution bottles.