Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Standard, 10,000 ppm Na, Ricca Chemical
SDP

Catalog No. 760516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7605-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7605-16 Supplier Ricca Chemical Company Supplier No. 760516
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A standard laboratory solution prepared for precision sodium analysis.

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Na
CAS Min % 96.0, 2.43
CAS Max % 99.0, 2.53
Equivalence 1 mL = 10 mg Na
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point Approximately 0°C
Boiling Point Approximately 100.1°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-element Standard Solution
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Sodium Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.