Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3

Catalog No. AC196311000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196311000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196311000 Supplier Thermo Scientific Chemicals Supplier No. 196311000
  • $69.80
    $56.65 / Each of 1

    Save $13.15

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
 Restricted Item
Add to Cart

CAS: 7631-99-4 | NNaO3 | 84.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium standard solution
Name Note 1 mg/ml Na in 0.5N HNO3
Packaging Plastic bottle
CAS 7631-99-4, 7697-37-2, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Liquid
Molecular Formula NNaO3
Concentration 997 to 1003 μg/mL of Na (1+) in 0.5N HNO3 (20°C)
MDL Number MFCD00011119
Color Colorless
Fieser 12,126; 16,303
Melting Point -3.0°C
Merck Index 15, 8778
Boiling Point 101.0°C
Solubility Soluble in water
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
Quantity 100 mL
PubChem CID 24268
ChEBI CHEBI:63005
Formula Weight 84.99
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.