Learn More
U.S. Pharmacopeia Sodium Starch Glycolate Type A, 9063-38-1, MFCD00217761, 400mg
Supplier:  U.S. Pharmacopeia 1614669
Encompass
Molecular formula (C2H4O3)x . (Na)x, Melting Point: Chars at about 200 ° C
Catalog No. NC0419341