Learn More
Aqua Solutions Sodium AA STD 1,000 mg/L NIST Traceable (in 0.5 mol/L HNO₃) (100ML)
SDP
Catalog No. NC9923697
Encompass_Preferred
Sodium AA STD 1,000 mg/L NIST Traceable (in 0.5 mol/L HNO₃) (100ML)
Catalog No. NC9923697 Supplier Aqua Solutions Supplier No. 8750100ML
Sodium AA STD 1,000 mg/L NIST Traceable (in 0.5 mol/L HNO₃) (100ML)